Assay Details
Gene Symbol: nsun4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007929.1 | NP_001007930.1 | 1 - 2 | 105 | 85 | |
GenBank mRNA | BC080376.1 | - | 1 - 2 | 105 | 85 | |
CR848182.2 | - | 1 - 2 | 105 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
