Assay Details
Gene Symbol: dctpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007936.2 | NP_001007937.2 | 3 - 4 | 236 | 70 | |
XM_012970323.2 | XP_012825777.1 | 3 - 4 | 248 | 70 | ||
XM_012970324.2 | XP_012825778.1 | 3 - 4 | 218 | 70 | ||
XM_012970325.2 | XP_012825779.1 | 3 - 4 | 273 | 70 | ||
GenBank mRNA | AL861133.2 | - | 3 - 4 | 215 | 70 | |
BC080443.1 | - | 3 - 4 | 253 | 70 | ||
CN113766.1 | - | 3 - 4 | 237 | 70 | ||
DN065473.1 | - | 2 - 3 | 165 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
