Assay Details
| Gene Symbol: amy2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007947.1 | NP_001007948.1 | 7 - 8 | 1137 | 88 | |
| GenBank mRNA | BC080454.1 | - | 7 - 8 | 1137 | 88 | |
| CR761580.2 | - | 7 - 8 | 1156 | 88 | ||
More Information
Set Membership: |
|

amy2b
Genomic Map