Assay Details
| Gene Symbol: ube2z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007968.2 | NP_001007969.2 | 3 - 4 | 542 | 72 | |
| GenBank mRNA | BC080480.1 | - | 3 - 4 | 543 | 72 | |
More Information
Set Membership: |
|

ube2z
Genomic Map