Assay Details
Gene Symbol: nabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007989.1 | NP_001007990.1 | 4 - 5 | 495 | 91 | |
GenBank mRNA | BC080502.1 | - | 4 - 5 | 495 | 91 | |
CR762209.2 | - | 4 - 5 | 418 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
