Assay Details
| Gene Symbol: sat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007996.1 | NP_001007997.1 | 3 - 4 | 362 | 87 | |
| XM_018091191.1 | XP_017946680.1 | 3 - 4 | 301 | 87 | ||
| GenBank mRNA | BC080875.1 | - | 3 - 4 | 362 | 87 | |
| CR942382.1 | - | 3 - 4 | 339 | 87 | ||
More Information
Set Membership: |
|

sat1
Genomic Map