Assay Details
Gene Symbol: rab7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008025.1 | NP_001008026.1 | 2 - 3 | 170 | 69 | |
GenBank mRNA | BC080905.1 | - | 2 - 3 | 170 | 69 | |
CR926163.1 | - | 2 - 3 | 145 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
