Assay Details
Gene Symbol: ppm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008029.1 | NP_001008030.1 | 3 - 4 | 1242 | 92 | |
XM_012962918.2 | XP_012818372.1 | 3 - 4 | 1565 | 92 | ||
XM_012962919.2 | XP_012818373.1 | 3 - 4 | 1304 | 92 | ||
XM_012962921.2 | XP_012818375.1 | 3 - 4 | 1563 | 92 | ||
XM_018093856.1 | XP_017949345.1 | 4 - 5 | 1620 | 92 | ||
XM_018093857.1 | XP_017949346.1 | 3 - 4 | 1563 | 92 | ||
GenBank mRNA | BC080911.1 | - | 3 - 4 | 1242 | 92 | |
CR761824.2 | - | 3 - 4 | 1094 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
