Assay Details
| Gene Symbol: blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008040.2 | NP_001008041.2 | 5 - 6 | 917 | 66 | |
| GenBank mRNA | BC080924.1 | - | 5 - 6 | 886 | 66 | |
| CR855426.2 | - | 5 - 6 | 867 | 66 | ||
More Information
Set Membership: |
|

blzf1
Genomic Map