Assay Details
| Gene Symbol: kdelr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008051.1 | NP_001008052.1 | 3 - 4 | 685 | 104 | |
| GenBank mRNA | BC080938.1 | - | 3 - 4 | 685 | 104 | |
| CR942706.2 | - | 3 - 4 | 387 | 104 | ||
More Information
Set Membership: |
|

kdelr3
Genomic Map