Assay Details
| Gene Symbol: trappc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008075.1 | NP_001008076.1 | 4 - 5 | 333 | 72 | |
| GenBank mRNA | BC080970.1 | - | 4 - 5 | 333 | 72 | |
| CR760179.2 | - | 4 - 5 | 357 | 72 | ||
More Information
Set Membership: |
|

trappc1
Genomic Map