Assay Details
Gene Symbol: chst13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008095.1 | NP_001008096.1 | 2 - 3 | 259 | 70 | |
GenBank mRNA | BC081295.1 | - | 2 - 3 | 259 | 70 | |
CR761652.2 | - | 2 - 3 | 275 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
