Assay Details
Gene Symbol: cidec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008101.1 | NP_001008102.1 | 4 - 5 | 385 | 67 | |
XM_012960954.2 | XP_012816408.1 | 3 - 4 | 929 | 67 | ||
XM_012960955.2 | XP_012816409.1 | 5 - 6 | 684 | 67 | ||
XM_012960956.2 | XP_012816410.1 | 3 - 4 | 373 | 67 | ||
XM_012960957.2 | XP_012816411.1 | 3 - 4 | 358 | 67 | ||
XM_018093023.1 | XP_017948512.1 | 4 - 5 | 582 | 67 | ||
GenBank mRNA | BC081304.1 | - | 4 - 5 | 385 | 67 | |
CR761715.2 | - | 4 - 5 | 393 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
