Assay Details
Gene Symbol: pigm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008119.1 | NP_001008120.1 | 471 | 78 | ||
GenBank mRNA | BC081324.1 | - | 471 | 78 | ||
CR760728.2 | - | 392 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
