Assay Details
Gene Symbol: nup35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008123.1 | NP_001008124.1 | 9 - 10 | 996 | 97 | |
XM_012970344.2 | XP_012825798.1 | 9 - 10 | 1092 | 97 | ||
XM_012970345.2 | XP_012825799.1 | 8 - 9 | 1046 | 97 | ||
XM_012970346.1 | XP_012825800.1 | 8 - 9 | 991 | 97 | ||
GenBank mRNA | BC081329.1 | - | 9 - 10 | 996 | 97 | |
CR761020.2 | - | 8 - 9 | 893 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
