Assay Details
Gene Symbol: larp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008143.2 | NP_001008144.1 | 1 - 2 | 366 | 78 | |
GenBank mRNA | BC081356.1 | - | 1 - 2 | 367 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
