Assay Details
Gene Symbol: larp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008143.2 | NP_001008144.1 | 2 - 3 | 584 | 101 | |
XM_012970348.2 | XP_012825802.1 | 2 - 3 | 1350 | 101 | ||
GenBank mRNA | BC081356.1 | - | 2 - 3 | 585 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
