Assay Details
| Gene Symbol: serpinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008152.1 | NP_001008153.1 | 5 - 6 | 1237 | 65 | |
| GenBank mRNA | BC081366.1 | - | 5 - 6 | 1237 | 65 | |
More Information
Set Membership: |
|

serpinc1
Genomic Map