Assay Details
Gene Symbol: ywhae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008155.1 | NP_001008156.1 | 1 - 2 | 237 | 87 | |
XM_012956823.1 | XP_012812277.1 | 1 - 2 | 304 | 87 | ||
GenBank mRNA | BC081369.1 | - | 1 - 2 | 237 | 87 | |
CR848241.2 | - | 1 - 2 | 97 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
