Assay Details
Gene Symbol: dyrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008157.1 | NP_001008158.1 | 2 - 3 | 275 | 74 | |
XM_012960516.2 | XP_012815970.1 | 3 - 4 | 783 | 74 | ||
GenBank mRNA | BC081371.1 | - | 2 - 3 | 275 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
