Assay Details
| Gene Symbol: txn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008160.1 | NP_001008161.1 | 3 - 4 | 459 | 96 | |
| XM_012960964.1 | XP_012816418.1 | 2 - 3 | 398 | 96 | ||
| GenBank mRNA | BC082341.1 | - | 3 - 4 | 459 | 96 | |
| CR761171.2 | - | 2 - 3 | 415 | 96 | ||
| CR762107.2 | - | 2 - 3 | 244 | 96 | ||
| CU075362.1 | - | 2 - 3 | 404 | 96 | ||
More Information
Set Membership: |
|

txn2
Genomic Map