Assay Details
Gene Symbol: nol12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008167.1 | NP_001008168.1 | 6 - 7 | 551 | 97 | |
XM_012960965.2 | XP_012816419.1 | 6 - 7 | 629 | 97 | ||
XM_012960966.2 | XP_012816420.1 | 6 - 7 | 640 | 97 | ||
XM_012960967.1 | XP_012816421.1 | 6 - 7 | 665 | 97 | ||
XM_012960968.2 | XP_012816422.1 | 6 - 7 | 634 | 97 | ||
GenBank mRNA | BC082486.1 | - | 6 - 7 | 551 | 97 | |
CR942728.2 | - | 6 - 7 | 552 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
