Assay Details
| Gene Symbol: adprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008181.1 | NP_001008182.2 | 456 | 103 | ||
| GenBank mRNA | BC082505.1 | - | 456 | 103 | ||
| CR855791.2 | - | 413 | 103 | |||
More Information
Set Membership: |
|

adprh
Genomic Map