Assay Details
| Gene Symbol: anxa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008182.1 | NP_001008183.1 | 9 - 10 | 688 | 67 | |
| GenBank mRNA | BC082506.1 | - | 9 - 10 | 688 | 67 | |
| CR761578.2 | - | 8 - 9 | 654 | 67 | ||
More Information
Set Membership: |
|

anxa5
Genomic Map