Assay Details
| Gene Symbol: bambi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008193.1 | NP_001008194.1 | 2 - 3 | 587 | 80 | |
| GenBank mRNA | BC082522.1 | - | 2 - 3 | 587 | 80 | |
| CR760560.2 | - | 2 - 3 | 564 | 80 | ||
More Information
Set Membership: |
|

bambi
Genomic Map