Assay Details
| Gene Symbol: tecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008198.2 | NP_001008199.1 | 5 - 6 | 339 | 74 | |
| XM_012953128.2 | XP_012808582.1 | 4 - 5 | 470 | 74 | ||
| GenBank mRNA | BC080144.1 | - | 5 - 6 | 340 | 74 | |
| BC091002.1 | - | 4 - 5 | 643 | 74 | ||
More Information
Set Membership: |
|

tecr
Genomic Map