Assay Details
Gene Symbol: cdc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008201.1 | NP_001008202.1 | 5 - 6 | 726 | 62 | |
GenBank mRNA | BC080871.1 | - | 5 - 6 | 726 | 62 | |
CR760436.2 | - | 5 - 6 | 639 | 62 | ||
CU464100.1 | - | 5 - 6 | 641 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
