Assay Details
| Gene Symbol: rpf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008430.3 | NP_001008430.2 | 9 - 10 | 829 | 76 | |
| XM_012962908.2 | XP_012818362.1 | 9 - 10 | 1013 | 76 | ||
| XM_018093849.1 | XP_017949338.1 | 7 - 8 | 880 | 76 | ||
| GenBank mRNA | BC080134.1 | - | 9 - 10 | 818 | 76 | |
| CX365896.2 | - | 1 - 2 | 153 | 76 | ||
More Information
Set Membership: |
|

rpf2
Genomic Map