Assay Details
Gene Symbol: gpat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008432.2 | NP_001008432.1 | 2 - 3 | 362 | 90 | |
GenBank mRNA | BC080141.1 | - | 2 - 3 | 364 | 90 | |
CR762294.2 | - | 2 - 3 | 307 | 90 |
Gene Symbol: gpat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR942493.2 | - | 3100 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
