Assay Details
Gene Symbol: gpat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008432.2 | NP_001008432.1 | 3 - 4 | 523 | 101 | |
GenBank mRNA | BC080141.1 | - | 3 - 4 | 525 | 101 | |
CR762294.2 | - | 3 - 4 | 468 | 101 |
Gene Symbol: gpat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR942493.2 | - | 3261 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
