Assay Details
Gene Symbol: fzd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008449.1 | NP_001008449.1 | 1 | 1651 | 62 | |
GenBank mRNA | BC080489.1 | - | 1 | 1651 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
