Assay Details
Gene Symbol: wdr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011071.1 | NP_001011071.1 | 2 - 3 | 252 | 54 | |
XM_012953486.2 | XP_012808940.1 | 2 - 3 | 233 | 54 | ||
GenBank mRNA | BC084454.1 | - | 2 - 3 | 252 | 54 | |
CR761781.2 | - | 1 - 2 | 160 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
