Assay Details
Gene Symbol: fbxl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011072.1 | NP_001011072.1 | 1 - 2 | 182 | 76 | |
XM_012960653.2 | XP_012816107.1 | 1 - 2 | 279 | 76 | ||
GenBank mRNA | BC084456.1 | - | 1 - 2 | 182 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
