Assay Details
Gene Symbol: akr1c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011079.1 | NP_001011079.1 | 559 | 82 | ||
GenBank mRNA | BC084466.1 | - | 559 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
