Assay Details
Gene Symbol: cecr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011089.1 | NP_001011089.1 | 788 | 92 | ||
GenBank mRNA | BC084481.1 | - | 788 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
