Assay Details
| Gene Symbol: rdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011091.1 | NP_001011091.1 | 5 - 6 | 1238 | 70 | |
| GenBank mRNA | BC084483.1 | - | 5 - 6 | 1238 | 70 | |
| CT030281.1 | - | 5 - 6 | 1233 | 70 | ||
More Information
Set Membership: |
|

rdh10
Genomic Map