Assay Details
Gene Symbol: cenpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011097.1 | NP_001011097.1 | 2 - 3 | 276 | 73 | |
XM_012960977.2 | XP_012816431.1 | 2 - 3 | 284 | 73 | ||
GenBank mRNA | BC084491.1 | - | 2 - 3 | 276 | 73 | |
CR848458.2 | - | 2 - 3 | 250 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
