Assay Details
Gene Symbol: nutf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011126.1 | NP_001011126.1 | 2 - 3 | 137 | 71 | |
GenBank mRNA | BC084526.1 | - | 2 - 3 | 137 | 71 | |
CR760342.2 | - | 1 - 2 | 126 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
