Assay Details
| Gene Symbol: eif4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011139.1 | NP_001011139.1 | 3 - 4 | 352 | 81 | |
| GenBank mRNA | BC084468.1 | - | 3 - 4 | 352 | 81 | |
| CR855769.2 | - | 3 - 4 | 351 | 81 | ||
More Information
Set Membership: |
|

eif4a1
Genomic Map