Assay Details
Gene Symbol: uap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011142.1 | NP_001011142.1 | 4 - 5 | 868 | 66 | |
XM_012960988.1 | XP_012816442.1 | 4 - 5 | 1534 | 66 | ||
GenBank mRNA | BC084497.1 | - | 4 - 5 | 868 | 66 | |
CR848106.2 | - | 4 - 5 | 864 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
