Assay Details
| Gene Symbol: ppp4r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011151.1 | NP_001011151.1 | 8 - 9 | 1078 | 71 | |
| GenBank mRNA | BC084901.1 | - | 8 - 9 | 1078 | 71 | |
| CR760404.1 | - | 8 - 9 | 964 | 71 | ||
More Information
Set Membership: |
|

ppp4r2
Genomic Map