Assay Details
Gene Symbol: fgfrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011189.1 | NP_001011189.1 | 5 - 6 | 1042 | 68 | |
GenBank mRNA | AJ616852.2 | - | 5 - 6 | 1042 | 68 | |
BC161343.1 | - | 5 - 6 | 1072 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
