Assay Details
| Gene Symbol: fgfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011189.1 | NP_001011189.1 | 6 - 7 | 1395 | 68 | |
| GenBank mRNA | AJ616852.2 | - | 6 - 7 | 1395 | 68 | |
| BC161343.1 | - | 6 - 7 | 1425 | 68 | ||
More Information
Set Membership: |
|

fgfrl1
Genomic Map