Assay Details
| Gene Symbol: cbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011190.1 | NP_001011190.1 | 2 - 3 | 446 | 99 | |
| GenBank mRNA | BC086506.2 | - | 2 - 3 | 446 | 99 | |
More Information
Set Membership: |
|

cbr1
Genomic Map