Assay Details
| Gene Symbol: bhmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011217.1 | NP_001011217.1 | 6 - 7 | 805 | 72 | |
| GenBank mRNA | BC087774.1 | - | 6 - 7 | 805 | 72 | |
| BC089235.1 | - | 6 - 7 | 789 | 72 | ||
More Information
Set Membership: |
|

bhmt
Genomic Map