Assay Details
| Gene Symbol: ube2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011221.1 | NP_001011221.1 | 6 - 7 | 471 | 65 | |
| XM_012970363.2 | XP_012825817.1 | 6 - 7 | 540 | 65 | ||
| GenBank mRNA | BC087785.1 | - | 6 - 7 | 471 | 65 | |
| CR848249.2 | - | 6 - 7 | 481 | 65 | ||
More Information
Set Membership: |
|

ube2f
Genomic Map