Assay Details
Gene Symbol: krt55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011224.1 | NP_001011224.1 | 3 - 4 | 841 | 97 | |
GenBank mRNA | BC087788.1 | - | 3 - 4 | 841 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
