Assay Details
| Gene Symbol: clec10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011225.1 | NP_001011225.1 | 4 - 5 | 314 | 75 | |
| XM_012960232.1 | XP_012815686.1 | 4 - 5 | 406 | 75 | ||
| XM_012960233.1 | XP_012815687.1 | 4 - 5 | 391 | 75 | ||
| XM_012960234.2 | XP_012815688.1 | 4 - 5 | 402 | 75 | ||
| XM_012960235.1 | XP_012815689.1 | 4 - 5 | 406 | 75 | ||
| GenBank mRNA | BC087789.1 | - | 4 - 5 | 314 | 75 | |
More Information
Set Membership: |
|

clec10a
Genomic Map