Assay Details
Gene Symbol: colec10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011227.1 | NP_001011227.1 | 4 - 5 | 400 | 63 | |
GenBank mRNA | BC087791.1 | - | 4 - 5 | 400 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
