Assay Details
Gene Symbol: rp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011242.1 | NP_001011242.1 | 4 - 5 | 413 | 65 | |
XM_012964601.2 | XP_012820055.1 | 4 - 5 | 579 | 65 | ||
XM_018094629.1 | XP_017950118.1 | 4 - 5 | 579 | 65 | ||
XM_018094630.1 | XP_017950119.1 | 4 - 5 | 467 | 65 | ||
XM_018094631.1 | XP_017950120.1 | 4 - 5 | 472 | 65 | ||
GenBank mRNA | BC087814.1 | - | 4 - 5 | 413 | 65 | |
CR761073.2 | - | 4 - 5 | 400 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
